This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01095539_m1
See other MMP16 GE Assays ›
Gene Symbol:
MMP16
Gene Name:
matrix metallopeptidase 16
Gene Aliases:
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP
Chromosome Location:
Chr.8: 88037232 - 88327488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01095539_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MMP16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005941.4 NP_005932.2 5 - 6 1151 120
GenBank mRNA AK314274.1 - 5 - 6 1151 120
BC069500.1 - 5 - 6 938 7262415 120
BC075004.2 - 5 - 6 895 30915400 120
BC075005.2 - 5 - 6 895 30915613 120
D83646.1 - 5 - 6 869 120
D83647.1 - 5 - 6 869 120
D85511.1 - 5 - 6 904 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800011041.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001396.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MMP16

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

endochondral ossification
proteolysis
extracellular matrix disassembly
collagen catabolic process
chondrocyte proliferation
positive regulation of catalytic activity
embryonic cranial skeleton morphogenesis
craniofacial suture morphogenesis
metalloendopeptidase activity
calcium ion binding
enzyme activator activity
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08018072.hg.1 ENST00000286614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08018072.hg.1 ENST00000544227.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08018072.hg.1 NM_005941.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08018072.hg.1 TC08001396.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0800175043.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay PSR0800175043.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800175043.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay PSR0800175043.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay PSR0800175043.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800175043.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay PSR0800175043.hg.1 TC0800011041.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000286614.10 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000520568.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000522726.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 ENST00000544227.5 Ensembl
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq
Affymetrix Human Clariom D Assay TC0800011041.hg.1 NM_005941.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0