This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01093485_s1
See other GEN1 GE Assays ›
Gene Symbol:
GEN1
Gene Name:
GEN1, Holliday junction 5' flap endonuclease
Gene Aliases:
Gen
Chromosome Location:
Chr.2: 17753687 - 17785920 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01093485_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GEN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC038243.1 - 1 62 116
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA828385.1 - 1 187 Chr. 2 - 17753772 - 17754087 [+] on Build GRCh38 116
GenBankmRNA CR617603.1 - 1 661 Chr. 2 - 17753296 - 17755047 [+] on Build GRCh38 116
GenBankmRNA BX427640.2 - 1 664 Chr. 2 - 17753296 - 17754252 [+] on Build GRCh38 116

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream structural maintenance of chromosomes 6 SMC6 SMC-6,
SMC6L1,
hSMC6
Anti-sense Chr. 2 - 17663812 - 17753831 on NCBI Build GRCh38
Downstream mesogenin 1 MSGN1 MSOG 61.5kb Sense Chr. 2 - 17816519 - 17817100 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000532257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000532257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000532257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000317402.11 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000381254.6 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000524465.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000528873.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000532257.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000532257.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000532257.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534451.2 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000534669.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 ENST00000614478.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_001130009.2 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq
Affymetrix Human Clariom D Assay TC0200006833.hg.1 NM_182625.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0