This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091453_s1
See other C18ORF12 GE Assays ›
Gene Symbol:
C18orf12
Gene Name:
chromosome 18 open reading frame 12
Gene Aliases:
HEIL1, HsT2508
Chromosome Location:
Chr.18: 48252301 - 48253185 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091453_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C18orf12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017026109.1 XP_016881598.1 1 247 65
GenBank mRNA AB027121.1 - 1 247 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger and BTB domain containing 7C ZBTB7C APM-1,
APM1,
ZBTB36,
ZNF857C
Anti-sense Chr. 18 - 48026886 - 48412639 on NCBI Build GRCh38
Downstream cap binding complex dependent translation initiation factor CTIF Gm672,
KIAA0427
286.2kb Sense Chr. 18 - 48539056 - 48863217 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800007265.hg.1 ENST00000623329.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012442.hg.1 ENST00000525475.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012442.hg.1 ENST00000525475.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007265.hg.1 ENST00000623329.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 207732 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0