This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01081092_s1
See other LOC286083 GE Assays ›
Gene Symbol:
LOC286083
Gene Name:
uncharacterized LOC286083
Gene Aliases:
-
Chromosome Location:
Chr.8: 1296034 - 1302607 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01081092_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC286083
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_111948.1 1 647 131
GenBank mRNA BC030648.2 - 1 647 131

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ERICH1 antisense RNA 1 ERICH1-AS1 C8orf68 -162.1kb Anti-sense Chr. 8 - 737605 - 1137777 on NCBI Build GRCh38
Downstream uncharacterized LOC105377776 LOC105377776 66.0kb Sense Chr. 8 - 1368648 - 1373228 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800009337.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911446 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184334 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184286 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911431 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184276 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911441 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184367 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911432 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184359 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911443 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184288 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184364 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911373 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184273 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184395 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184272 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911355 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911453 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911376 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911427 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184380 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184302 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16054150 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184385 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911448 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16054125 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184396 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911468 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184345 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184407 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184342 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911466 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911463 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184331 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184358 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184399 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184322 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911375 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911439 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184321 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184306 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184296 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184403 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911422 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184320 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911339 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184405 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184315 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184311 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911359 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184300 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911382 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184402 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911406 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911390 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184351 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184339 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184327 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911455 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911330 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184328 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184298 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184369 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911416 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911461 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911353 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184307 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184374 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184293 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911366 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911331 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184285 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16054130 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911393 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911371 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911372 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184325 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911429 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911397 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911458 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184363 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911434 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184310 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184387 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911367 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911345 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911449 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911380 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911437 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911410 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911438 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184408 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911349 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911404 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911392 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911415 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911457 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184292 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16054127 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184370 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184376 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911419 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911401 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911409 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911426 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26184291 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P05911333 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AB266487.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AF182420.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AK022802.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AK092614.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AK093635.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AK310279.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AK315650.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 AX748234.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 BC009683.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 BC024268.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 CR456472.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 CR611013.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 CR619121.1 genbank_mrna_human
Agilent SurePrint G3 Human CGH Microarray 4x180K 132276 NM_021822.1 refseq_mrna_human_NM
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 795146 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 526476 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3832532 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 1864 - -
Agilent Human Genome CGH + SNP Microarray A_16_P18189967 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3832532 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3832532 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18189967 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18189967 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 157558 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 102115 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38279275 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18189967 - -
Agilent Human 1B Microarray G4111A A_32_P377255 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18189967 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1561257_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164017.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164017.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164017.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164017.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164017.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164017.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164017.hg.1 TC0800009337.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay TC0800009337.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164015.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164015.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164015.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164015.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164015.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164015.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164015.hg.1 TC0800009337.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800164013.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164013.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164013.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164013.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164013.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164013.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164013.hg.1 TC0800009337.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800164018.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164018.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164018.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164018.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164018.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164018.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164018.hg.1 TC0800009337.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800164016.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164016.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164016.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164016.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164016.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164016.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164016.hg.1 TC0800009337.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800164014.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164014.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164014.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164014.hg.1 ENST00000521186.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164014.hg.1 ENST00000617093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800164014.hg.1 NR_111948.1 Refseq
Affymetrix Human Clariom D Assay PSR0800164014.hg.1 TC0800009337.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 1561257_at - -
Human Genome U133 Plus 2.0 Array 1561257_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561257_at9 - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A000034 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A000034 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 232506 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1840957 NR_033895.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1840957 BC030648.2 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1840957 NR_033895.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1840957 BC030648.2 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1840957 NR_033895.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1840957 BC030648.2 human_transcriptome
Illumina Sentrix Human-6 v2 Expression BeadChip 6960220 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_88925 - -
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1840957 NR_033895.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1840957 BC030648.2 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1840957 NR_033895.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1840957 BC030648.2 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1840957 NR_033895.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1840957 BC030648.2 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header