This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01080086_m1
See other SIPA1L3 GE Assays ›
Gene Symbol:
SIPA1L3
Gene Name:
signal induced proliferation associated 1 like 3
Gene Aliases:
CTRCT45, SPAL3, SPAR3
Chromosome Location:
Chr.19: 37906823 - 38208372 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01080086_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIPA1L3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015073.2 NP_055888.1 12 - 13 4053 95
XM_005258671.4 XP_005258728.1 12 - 13 4494 95
XM_011526657.2 XP_011524959.1 12 - 13 4848 95
XM_017026517.1 XP_016882006.1 13 - 14 4948 95
XM_017026518.1 XP_016882007.1 12 - 13 4450 95
GenBank mRNA AB011117.2 - 10 - 11 2299 95
AY168880.1 - 10 - 11 3799 95
BC150620.1 - 12 - 13 4032 9056990 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900007988.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000521.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SIPA1L3

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

eye development
hematopoietic progenitor cell differentiation
epithelial cell morphogenesis
cytoskeleton organization
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
establishment of epithelial cell polarity
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19006923.hg.1 ENST00000222345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19006923.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19006923.hg.1 TC19000521.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000476317.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000476317.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000476317.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595495.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595495.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595495.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595982.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000595982.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000599644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000599644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000600919.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000600919.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000600919.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601222.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601222.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601222.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000476317.2 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000476317.2 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000476317.2 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000594553.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595384.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595495.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595495.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595495.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595982.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000595982.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000596403.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000599644.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000599644.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000600919.2 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000600919.2 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000600919.2 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601054.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601222.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601222.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601222.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 ENST00000601881.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay TC1900007988.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay PSR1900159831.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900159831.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay PSR1900159831.hg.1 ENST00000222345.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900159831.hg.1 NM_015073.2 Refseq
Affymetrix Human Clariom D Assay PSR1900159831.hg.1 TC1900007988.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0