This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01079897_g1
See other UBE2Q1 GE Assays ›
Gene Symbol:
UBE2Q1
Gene Name:
ubiquitin conjugating enzyme E2 Q1
Gene Aliases:
GTAP, NICE-5, PRO3094, UBE2Q
Chromosome Location:
Chr.1: 154548575 - 154558644 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01079897_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2Q1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017582.6 NP_060052.3 10 - 11 1163 119
GenBank mRNA AF116721.1 - 3 - 4 139 119
AJ243666.1 - 4 - 5 230 119
AK056388.1 - 6 - 7 463 119
AK293986.1 - 10 - 11 786 119
AY112698.1 - 10 - 11 1138 119
AY948200.1 - 10 - 11 1146 119
BC000848.1 - 6 - 7 497 119
BC009286.2 - 5 - 6 301 119
BC015316.1 - 2 - 3 106 119
BC061583.1 - 6 - 7 475 119
BC070158.1 - 9 - 10 688 119
BC087836.1 - 8 - 9 574 119
BC107057.2 - 11 - 12 831 119
BC148185.2 - 10 - 11 840 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015918.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003293.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF116721.1,AJ243666.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UBE2Q1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

suckling behavior
embryo implantation
mating behavior
fertilization
protein ubiquitination
reproductive system development
prolactin secretion
protein binding
ATP binding
ubiquitin conjugating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01050786.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01050786.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01050786.hg.1 TC01003293.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01050787.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01050787.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01050787.hg.1 TC01003293.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000474181.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 ENST00000497453.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay TC0100015918.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay PSR0100219817.hg.1 TC0100015918.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000292211.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000467683.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000476612.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000483639.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 ENST00000491572.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 NM_017582.6 Refseq
Affymetrix Human Clariom D Assay PSR0100219818.hg.1 TC0100015918.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0