This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01077029_m1
See other CD247 GE Assays ›
Gene Symbol:
CD247
Gene Name:
CD247 molecule
Gene Aliases:
CD3-ZETA, CD3H, CD3Q, CD3Z, IMD25, T3Z, TCRZ
Chromosome Location:
Chr.1: 167430640 - 167518610 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01077029_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD247
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198053.2 NP_932170.1 5 - 6 478 70
XM_011510144.2 XP_011508446.1 6 - 7 546 70
XM_017002800.1 XP_016858289.1 6 - 7 537 70
GenBank mRNA BC025703.1 - 5 - 6 394 5222574 70
X55512.1 - 5 - 6 377 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD247

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

viral process
T cell costimulation
Fc-gamma receptor signaling pathway involved in phagocytosis
regulation of defense response to virus by virus
regulation of immune response
T cell receptor signaling pathway
transmembrane signaling receptor activity
protein binding
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0