This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01071053_m1
See other RASA3 GE Assays ›
Gene Symbol:
RASA3
Gene Name:
RAS p21 protein activator 3
Gene Aliases:
GAP1IP4BP, GAPIII
Chromosome Location:
Chr.13: 113977783 - 114132623 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01071053_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RASA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320821.1 NP_001307750.1 10 - 11 1273 79
NM_001320822.1 NP_001307751.1 8 - 9 795 79
NM_007368.3 NP_031394.2 8 - 9 807 79
XM_011534841.2 XP_011533143.1 8 - 9 872 79
XM_011534842.2 XP_011533144.1 8 - 9 932 79
XM_017020438.1 XP_016875927.1 10 - 11 1221 79
XM_017020439.1 XP_016875928.1 9 - 10 1018 79
GenBank mRNA AK128094.1 - 10 - 11 1242 79
AK289746.1 - 8 - 9 713 79
AK294913.1 - 8 - 9 780 79
AK295861.1 - 7 - 8 641 79
AK308880.1 - 5 - 6 869 79
AK313795.1 - 8 - 9 741 79
BC038456.1 - 8 - 9 804 5240667 79
BC047242.1 - 8 - 9 795 6042834 79
X89399.1 - 8 - 9 719 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009933.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000901.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001320821.1,XM_017020438.1,XM_017020439.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RASA3

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
signal transduction
positive regulation of GTPase activity
negative regulation of Ras protein signal transduction
release of sequestered calcium ion into cytosol
GTPase activator activity
calcium-release channel activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13010126.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13010126.hg.1 ENST00000389544.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13010126.hg.1 ENST00000542651.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13010126.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13010126.hg.1 TC13000901.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300166154.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300166154.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300166154.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay PSR1300166154.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300166154.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay PSR1300166154.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay PSR1300166154.hg.1 TC1300009933.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0