This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01071036_m1
See other RASA3 GE Assays ›
Gene Symbol:
RASA3
Gene Name:
RAS p21 protein activator 3
Gene Aliases:
GAP1IP4BP, GAPIII
Chromosome Location:
Chr.13: 113977783 - 114132623 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01071036_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RASA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320821.1 NP_001307750.1 16 - 17 2008 109
NM_001320822.1 NP_001307751.1 14 - 15 1530 109
NM_007368.3 NP_031394.2 14 - 15 1542 109
XM_011534841.2 XP_011533143.1 14 - 15 1607 109
XM_011534842.2 XP_011533144.1 14 - 15 1667 109
XM_017020438.1 XP_016875927.1 16 - 17 1956 109
XM_017020439.1 XP_016875928.1 15 - 16 1753 109
GenBank mRNA AK128094.1 - 16 - 17 1977 109
AK289746.1 - 14 - 15 1448 109
AK294913.1 - 14 - 15 1515 109
AK313795.1 - 14 - 15 1476 109
BC038456.1 - 14 - 15 1539 5240667 109
BC047242.1 - 14 - 15 1530 6042834 109
X89399.1 - 14 - 15 1454 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009933.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000901.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RASA3

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
signal transduction
positive regulation of GTPase activity
negative regulation of Ras protein signal transduction
release of sequestered calcium ion into cytosol
GTPase activator activity
calcium-release channel activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13010119.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13010119.hg.1 ENST00000389544.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13010119.hg.1 NM_007368.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13010119.hg.1 TC13000901.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300166147.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300166147.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay PSR1300166147.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300166147.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay PSR1300166147.hg.1 TC1300009933.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000334062.7 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 ENST00000542651.2 Ensembl
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq
Affymetrix Human Clariom D Assay TC1300009933.hg.1 NM_007368.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0