This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061437_m1
See other DDX58 GE Assays ›
Gene Symbol:
DDX58
Gene Name:
DEXD/H-box helicase 58
Gene Aliases:
RIG-I, RIGI, RLR-1, SGMRT2
Chromosome Location:
Chr.9: 32455302 - 32526324 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061437_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDX58
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014314.3 NP_055129.2 1 - 2 265 92
GenBank mRNA AF038963.1 - 1 - 2 264 92
AK301678.1 - 1 - 2 136 92
AK315040.1 - 1 - 2 136 92
BC107731.1 - 1 - 2 287 92
BC132786.1 - 1 - 2 213 40146808 92
BC136610.1 - 1 - 2 213 9020600 92
DN998145.1 - 1 - 2 266 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009825.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000999.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK301678.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DDX58

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
detection of virus
response to virus
positive regulation of gene expression
regulation of cell migration
negative regulation of type I interferon production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
regulation of type III interferon production
cytoplasmic pattern recognition receptor signaling pathway in response to virus
RIG-I signaling pathway
positive regulation of transcription factor import into nucleus
response to exogenous dsRNA
innate immune response
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
double-stranded DNA binding
double-stranded RNA binding
single-stranded RNA binding
helicase activity
protein binding
ATP binding
zinc ion binding
identical protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379868.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 ENST00000379883.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009825.hg.1 NM_014314.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 131033 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0