This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01061237_g1
See other NDRG4 GE Assays ›
Gene Symbol:
NDRG4
Gene Name:
NDRG family member 4
Gene Aliases:
BDM1, SMAP-8, SMAP8
Chromosome Location:
Chr.16: 58463645 - 58515403 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01061237_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDRG4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130487.1 NP_001123959.1 9 - 10 1012 89
NM_001242833.1 NP_001229762.1 8 - 9 711 89
NM_001242834.1 NP_001229763.1 7 - 8 675 89
NM_001242835.1 NP_001229764.1 7 - 8 621 89
NM_001242836.1 NP_001229765.1 7 - 8 621 89
NM_020465.3 NP_065198.1 9 - 10 787 89
NM_022910.3 NP_075061.1 9 - 10 884 89
NR_040072.1 7 - 8 713 89
XM_006721252.3 XP_006721315.1 10 - 11 887 89
XM_006721253.3 XP_006721316.1 10 - 11 820 89
XM_006721254.3 XP_006721317.1 9 - 10 874 89
XM_006721256.3 XP_006721319.1 10 - 11 818 89
XM_006721257.3 XP_006721320.1 9 - 10 859 89
XM_006721258.3 XP_006721321.1 9 - 10 763 89
XM_006721259.3 XP_006721322.1 9 - 10 828 89
XM_006721260.3 XP_006721323.1 9 - 10 766 89
XM_006721261.3 XP_006721324.1 8 - 9 711 89
XM_006721262.3 XP_006721325.1 7 - 8 792 89
XM_011523290.2 XP_011521592.1 10 - 11 974 89
XM_011523291.2 XP_011521593.1 9 - 10 938 89
XM_011523292.2 XP_011521594.1 10 - 11 974 89
XM_011523293.2 XP_011521595.1 9 - 10 932 89
XM_011523294.2 XP_011521596.1 10 - 11 753 89
XM_017023582.1 XP_016879071.1 9 - 10 938 89
XM_017023583.1 XP_016879072.1 10 - 11 974 89
XM_017023584.1 XP_016879073.1 10 - 11 887 89
XM_017023585.1 XP_016879074.1 9 - 10 874 89
XM_017023586.1 XP_016879075.1 9 - 10 821 89
XM_017023587.1 XP_016879076.1 10 - 11 818 89
XM_017023588.1 XP_016879077.1 9 - 10 763 89
XM_017023589.1 XP_016879078.1 9 - 10 828 89
XM_017023590.1 XP_016879079.1 10 - 11 753 89
GenBank mRNA AB021172.1 - 9 - 10 687 89
AB033006.2 - 7 - 8 607 89
AB044944.1 - 7 - 8 621 89
AB044945.1 - 7 - 8 621 89
AB044946.1 - 8 - 9 634 89
AF308608.1 - 7 - 8 628 89
AK055038.1 - 7 - 8 623 89
AK124005.1 - 3 - 4 590 89
AK126574.1 - 9 - 10 784 89
AK126729.1 - 10 - 11 2734 89
AK131248.1 - 7 - 8 675 89
AK296415.1 - 7 - 8 675 89
AK299949.1 - 8 - 9 711 89
AK316089.1 - 7 - 8 713 89
AL136584.1 - 7 - 8 607 89
BC001527.1 - 7 - 8 605 89
BC011795.2 - 7 - 8 609 4139594 89
CR936733.1 - 9 - 10 698 89
KU178760.1 - 7 - 8 515 89
KU178761.1 - 7 - 8 515 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000503.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDRG4

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

heart looping
response to stress
brain development
visual learning
negative regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of neuron projection development
negative regulation of smooth muscle cell migration
cell growth
cell differentiation
embryonic heart tube development
vesicle docking
negative regulation of smooth muscle cell proliferation
cardiac muscle cell proliferation
cell migration involved in heart development
positive regulation of ERK1 and ERK2 cascade
regulation of endocytic recycling
molecular_function
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 ENST00000258187.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 ENST00000356752.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 ENST00000394279.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 ENST00000394282.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 ENST00000421602.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NM_001130487.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NM_001242833.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NM_001242834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NM_001242835.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NM_001242836.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NM_020465.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NM_022910.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 NR_040072.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16007073.hg.1 TC16000503.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0