This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01059742_g1
See other AGO4 GE Assays ›
Gene Symbol:
AGO4
Gene Name:
argonaute 4, RISC catalytic component
Gene Aliases:
EIF2C4
Chromosome Location:
Chr.1: 35807630 - 35857890 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01059742_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGO4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017629.3 NP_060099.2 9 - 10 1305 92
XM_005270578.3 XP_005270635.1 9 - 10 1190 92
XM_005270579.3 XP_005270636.1 8 - 9 998 92
XM_011540883.2 XP_011539185.1 6 - 7 794 92
XM_011540884.2 XP_011539186.1 6 - 7 657 92
GenBank mRNA AB046787.1 - 9 - 10 1305 92
AL831908.1 - 8 - 9 1092 92
BC152450.1 - 9 - 10 1305 8862582 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007784.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000462.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AGO4

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA catabolic process
Wnt signaling pathway, calcium modulating pathway
RNA secondary structure unwinding
pre-miRNA processing
posttranscriptional gene silencing by RNA
production of miRNAs involved in gene silencing by miRNA
miRNA mediated inhibition of translation
mRNA cleavage involved in gene silencing by miRNA
miRNA loading onto RISC involved in gene silencing by miRNA
phosphatidylinositol-mediated signaling
mRNA cleavage involved in gene silencing by siRNA
double-stranded RNA binding
single-stranded RNA binding
protein binding
miRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01007111.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007111.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007111.hg.1 TC01000462.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000496257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000496257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 ENST00000496257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000488778.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000496257.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000496257.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 ENST00000496257.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay TC0100007784.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154778.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100154778.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154778.hg.1 ENST00000373210.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100154778.hg.1 NM_017629.3 Refseq
Affymetrix Human Clariom D Assay PSR0100154778.hg.1 TC0100007784.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0