This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01053423_m1
See other UBE2D1 GE Assays ›
Gene Symbol:
UBE2D1
Gene Name:
ubiquitin conjugating enzyme E2 D1
Gene Aliases:
E2(17)KB1, SFT, UBC4/5, UBCH5, UBCH5A
Chromosome Location:
Chr.10: 58334979 - 58370753 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01053423_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2D1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204880.1 NP_001191809.1 4 - 5 498 82
NM_003338.4 NP_003329.1 5 - 6 530 82
XM_017016609.1 XP_016872098.1 5 - 6 429 82
GenBank mRNA AF257220.1 - 5 - 6 453 82
AJ272367.1 - 5 - 6 453 82
AK291901.1 - 5 - 6 501 82
BC005980.1 - 5 - 6 501 4106710 82
BC015997.1 - 5 - 6 505 4305900 82
BI598342.1 - 5 - 6 534 82
BT007041.1 - 5 - 6 307 82
DA618122.1 - 4 - 5 471 82
DA677507.1 - 5 - 6 530 82
X78140.1 - 5 - 6 322 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000007700.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000360.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UBE2D1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
ubiquitin-dependent protein catabolic process
BMP signaling pathway
anaphase-promoting complex-dependent catabolic process
positive regulation of protein ubiquitination
TRIF-dependent toll-like receptor signaling pathway
Fc-epsilon receptor signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
regulation of transcription from RNA polymerase II promoter in response to hypoxia
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
ATP binding
ubiquitin conjugating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10004154.hg.1 ENST00000373910.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10004154.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10004154.hg.1 NM_001204880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10004154.hg.1 NM_003338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10004154.hg.1 TC10000360.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay PSR1000152311.hg.1 TC1000007700.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000373910.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000473824.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000606483.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 ENST00000615793.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_001204880.1 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq
Affymetrix Human Clariom D Assay TC1000007700.hg.1 NM_003338.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0