Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.1 ST Array |
8083318 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
237116_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014551.hg.1 |
NR_036538.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014551.hg.1 |
TC03000810.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300165620.hg.1 |
NR_036538.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165620.hg.1 |
NR_036538.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165620.hg.1 |
TC0300009149.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
237116_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
237116_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
237116_at2 |
- |
- |
Human Genome U95 Set |
78100_PM_at |
- |
- |
Human Genome U95 Set |
78100_at |
- |
- |
Human Genome U133 Set |
237116_PM_at |
- |
- |
Human Genome U133 Set |
237116_at |
- |
- |