This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01052520_g1
See other KRT3 GE Assays ›
Gene Symbol:
KRT3
Gene Name:
keratin 3
Gene Aliases:
CK3, K3
Chromosome Location:
Chr.12: 52789685 - 52796108 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01052520_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KRT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_057088.2 NP_476429.2 4 - 5 1093 89
GenBank mRNA AJ628418.1 - 4 - 5 982 89
AK314987.1 - 4 - 5 1102 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200010775.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001535.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KRT3

Gene Ontology Categories:

Function(s) Process(es)

epithelial cell differentiation
intermediate filament cytoskeleton organization
structural molecule activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12020164.hg.1 ENST00000309505.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12020164.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12020164.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12020164.hg.1 TC12001535.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12020165.hg.1 ENST00000309505.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12020165.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12020165.hg.1 NM_057088.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12020165.hg.1 TC12001535.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200182197.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200182197.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay PSR1200182197.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200182197.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay PSR1200182197.hg.1 TC1200010775.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay TC1200010775.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay PSR1200182198.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200182198.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay PSR1200182198.hg.1 ENST00000417996.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200182198.hg.1 NM_057088.2 Refseq
Affymetrix Human Clariom D Assay PSR1200182198.hg.1 TC1200010775.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0