This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01050988_m1
See other INO80 GE Assays ›
Gene Symbol:
INO80
Gene Name:
INO80 complex subunit
Gene Aliases:
INO80A, INOC1, hINO80
Chromosome Location:
Chr.15: 40978880 - 41117081 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01050988_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: INO80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017553.2 NP_060023.1 32 - 33 4230 99
NR_104038.1 31 - 32 4102 99
XM_011521686.2 XP_011519988.1 18 - 19 2041 99
GenBank mRNA AB033085.1 - 32 - 33 4126 99
AK002176.1 - 4 - 5 394 99
BC144681.1 - 31 - 32 3988 99
BC146785.1 - 32 - 33 4138 9007241 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009138.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001246.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene INO80

Gene Ontology Categories:

Function(s) Process(es)

mitotic sister chromatid segregation
double-strand break repair via homologous recombination
double-strand break repair
chromatin remodeling
transcription, DNA-templated
positive regulation of nuclear cell cycle DNA replication
positive regulation of cell growth
DNA duplex unwinding
cellular response to UV
positive regulation of transcription from RNA polymerase II promoter
spindle assembly
cell division
UV-damage excision repair
cellular response to ionizing radiation
regulation of G1/S transition of mitotic cell cycle
DNA binding
DNA helicase activity
actin binding
protein binding
ATP binding
ATPase activity
alpha-tubulin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15011267.hg.1 ENST00000263793.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15011267.hg.1 ENST00000361937.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15011267.hg.1 ENST00000401393.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15011267.hg.1 NM_017553.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15011267.hg.1 TC15001246.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000361937.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000401393.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000558357.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000616814.4 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 NM_017553.2 Refseq
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 NR_104038.1 Refseq
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000361937.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000401393.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000558357.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 ENST00000616814.4 Ensembl
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 NM_017553.2 Refseq
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 NR_104038.1 Refseq
Affymetrix Human Clariom D Assay PSR1500166160.hg.1 TC1500009138.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0