This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01038901_m1
See other PIKFYVE GE Assays ›
Gene Symbol:
PIKFYVE
Gene Name:
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases:
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location:
Chr.2: 208266178 - 208358751 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01038901_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIKFYVE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015040.3 NP_055855.2 22 - 23 3947 119
XM_011510778.2 XP_011509080.1 23 - 24 3997 119
XM_011510779.2 XP_011509081.1 23 - 24 3930 119
XM_011510780.2 XP_011509082.1 23 - 24 4015 119
XM_011510781.2 XP_011509083.1 23 - 24 3938 119
XM_011510782.2 XP_011509084.1 23 - 24 3997 119
XM_011510783.2 XP_011509085.1 22 - 23 3829 119
XM_011510784.2 XP_011509086.1 22 - 23 3847 119
XM_011510785.2 XP_011509087.1 22 - 23 3769 119
XM_011510786.2 XP_011509088.1 21 - 22 3664 119
XM_011510787.1 XP_011509089.1 21 - 22 3673 119
XM_011510788.1 XP_011509090.1 20 - 21 3656 119
XM_011510789.2 XP_011509091.1 19 - 20 3478 119
XM_011510790.1 XP_011509092.1 21 - 22 3775 119
XM_011510792.2 XP_011509094.1 23 - 24 3997 119
XM_017003568.1 XP_016859057.1 22 - 23 3833 119
XM_017003569.1 XP_016859058.1 21 - 22 3665 119
XM_017003570.1 XP_016859059.1 19 - 20 3514 119
XM_017003571.1 XP_016859060.1 18 - 19 3310 119
XM_017003572.1 XP_016859061.1 21 - 22 3651 119
XM_017003573.1 XP_016859062.1 18 - 19 3359 119
XM_017003574.1 XP_016859063.1 17 - 18 3030 119
GenBank mRNA AK127769.1 - 21 - 22 3684 119
AY457063.1 - 22 - 23 3951 119
EU794609.1 - 22 - 23 3951 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200010624.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001235.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PIKFYVE

Panther Classification:

Molecular Function -

kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylinositol biosynthetic process
receptor-mediated endocytosis
myelin assembly
protein localization to nucleus
intracellular signal transduction
phosphatidylinositol-3-phosphate biosynthetic process
retrograde transport, endosome to Golgi
phosphatidylinositol phosphorylation
phosphatidylinositol 5-phosphate metabolic process
regulation of autophagosome assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
zinc ion binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02018925.hg.1 ENST00000264380.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02018925.hg.1 ENST00000392200.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02018925.hg.1 NM_015040.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02018925.hg.1 TC02001235.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200175285.hg.1 ENST00000264380.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200175285.hg.1 ENST00000452564.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175285.hg.1 NM_015040.3 Refseq
Affymetrix Human Clariom D Assay PSR0200175285.hg.1 ENST00000264380.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200175285.hg.1 ENST00000452564.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200175285.hg.1 NM_015040.3 Refseq
Affymetrix Human Clariom D Assay PSR0200175285.hg.1 TC0200010624.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0