This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01037713_g1
See other PDK4 GE Assays ›
Gene Symbol:
PDK4
Gene Name:
pyruvate dehydrogenase kinase 4
Gene Aliases:
-
Chromosome Location:
Chr.7: 95583497 - 95596613 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01037713_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDK4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002612.3 NP_002603.1 8 - 9 1186 138
GenBank mRNA AK096428.1 - 9 - 10 985 138
AK098151.1 - 4 - 5 500 138
AK223328.1 - 8 - 9 1082 138
AK312424.1 - 8 - 9 1092 138
BC040239.1 - 8 - 9 1066 3047987 138
U54617.1 - 8 - 9 1088 138

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700011842.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001618.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDK4

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
glucose metabolic process
protein phosphorylation
regulation of pH
insulin receptor signaling pathway
cellular response to starvation
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of cellular ketone metabolic process
regulation of glucose metabolic process
regulation of fatty acid biosynthetic process
glucose homeostasis
response to starvation
regulation of bone resorption
regulation of fatty acid oxidation
cellular response to fatty acid
reactive oxygen species metabolic process
xenophagy
negative regulation of anoikis
protein kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07024624.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024624.hg.1 ENST00000542888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024624.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07024624.hg.1 TC07001618.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07024626.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024626.hg.1 ENST00000542888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024626.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07024626.hg.1 TC07001618.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07024625.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024625.hg.1 ENST00000542888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07024625.hg.1 NM_002612.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07024625.hg.1 TC07001618.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay PSR0700184657.hg.1 TC0700011842.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473301.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay TC0700011842.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay PSR0700184656.hg.1 TC0700011842.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000005178.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000468445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000473796.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 ENST00000498190.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 NM_002612.3 Refseq
Affymetrix Human Clariom D Assay PSR0700184658.hg.1 TC0700011842.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0