This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01031362_m1
See other MYCBP2 GE Assays ›
Gene Symbol:
MYCBP2
Gene Name:
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases:
Myc-bp2, PAM, Phr
Chromosome Location:
Chr.13: 77044657 - 77327066 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01031362_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYCBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015057.4 NP_055872.4 39 - 40 6163 123
XM_005266299.2 XP_005266356.1 39 - 40 6187 123
XM_006719779.2 XP_006719842.1 39 - 40 6187 123
XM_006719780.2 XP_006719843.1 39 - 40 6187 123
XM_006719781.2 XP_006719844.1 39 - 40 6187 123
XM_006719782.2 XP_006719845.1 39 - 40 6187 123
XM_006719783.2 XP_006719846.1 39 - 40 6187 123
XM_006719784.2 XP_006719847.1 38 - 39 6079 123
XM_006719785.2 XP_006719848.1 39 - 40 6187 123
XM_006719786.2 XP_006719849.1 39 - 40 6187 123
XM_006719787.2 XP_006719850.1 39 - 40 6187 123
XM_006719788.2 XP_006719851.1 39 - 40 6187 123
XM_006719789.2 XP_006719852.1 39 - 40 6187 123
XM_011535004.2 XP_011533306.1 38 - 39 5726 123
XM_011535005.2 XP_011533307.1 22 - 23 3267 123
XM_017020454.1 XP_016875943.1 39 - 40 6187 123
XM_017020455.1 XP_016875944.1 39 - 40 6187 123
XM_017020456.1 XP_016875945.1 39 - 40 6187 123
XM_017020457.1 XP_016875946.1 39 - 40 6187 123
XM_017020458.1 XP_016875947.1 38 - 39 6079 123
XM_017020459.1 XP_016875948.1 38 - 39 6079 123
XM_017020460.1 XP_016875949.1 39 - 40 6187 123
XM_017020461.1 XP_016875950.1 39 - 40 6187 123
XM_017020462.1 XP_016875951.1 39 - 40 6187 123
XM_017020463.1 XP_016875952.1 38 - 39 6079 123
XM_017020464.1 XP_016875953.1 39 - 40 6187 123
XM_017020465.1 XP_016875954.1 38 - 39 6079 123
XM_017020466.1 XP_016875955.1 39 - 40 6187 123
XM_017020467.1 XP_016875956.1 39 - 40 6187 123
XM_017020468.1 XP_016875957.1 38 - 39 5674 123
GenBank mRNA AF075587.1 - 40 - 41 5928 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009293.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000744.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MYCBP2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
protein ubiquitination
protein binding
zinc ion binding
ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13008608.hg.1 ENST00000357337.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008608.hg.1 ENST00000360084.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008608.hg.1 ENST00000407578.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008608.hg.1 ENST00000544440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008608.hg.1 NM_015057.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13008608.hg.1 TC13000744.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300162415.hg.1 ENST00000357337.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300162415.hg.1 ENST00000544440.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300162415.hg.1 NM_015057.4 Refseq
Affymetrix Human Clariom D Assay PSR1300162415.hg.1 ENST00000357337.10 Ensembl
Affymetrix Human Clariom D Assay PSR1300162415.hg.1 ENST00000544440.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300162415.hg.1 NM_015057.4 Refseq
Affymetrix Human Clariom D Assay PSR1300162415.hg.1 TC1300009293.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0