This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01029890_g1
See other TAF7L GE Assays ›
Gene Symbol:
TAF7L
Gene Name:
TATA-box binding protein associated factor 7 like
Gene Aliases:
CT40, TAF2Q
Chromosome Location:
Chr.X: 101268253 - 101293071 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01029890_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAF7L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168474.1 NP_001161946.1 5 - 6 523 120
NM_024885.3 NP_079161.3 5 - 6 691 120
XM_005262152.1 XP_005262209.1 5 - 6 523 120
XM_006724664.1 XP_006724727.1 5 - 6 690 120
GenBank mRNA AF285595.1 - 5 - 6 525 120
AK026810.1 - 4 - 5 669 120
AK308261.1 - 5 - 6 693 121
BC043391.1 - 5 - 6 677 6154641 120
DB042169.1 - 5 - 6 523 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00010351.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001215.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAF7L

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
biological_process
cell differentiation
RNA polymerase II transcriptional preinitiation complex assembly
molecular_function
transcription coactivator activity
transcription factor binding
histone acetyltransferase binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X016356.hg.1 ENST00000324762.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X016356.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X016356.hg.1 ENST00000372905.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X016356.hg.1 ENST00000372907.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X016356.hg.1 NM_001168474.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X016356.hg.1 NM_024885.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X016356.hg.1 TC0X001215.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay TC0X00010351.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 ENST00000324762.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 ENST00000356784.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 ENST00000372907.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 NM_001168474.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 NM_024885.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00170339.hg.1 TC0X00010351.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0