This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01025719_m1
See other ATE1 GE Assays ›
Gene Symbol:
ATE1
Gene Name:
arginyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 121740421 - 121928801 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01025719_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001288735.1 NP_001275664.1 5 - 6 934 60
NM_001288736.1 NP_001275665.1 7 - 8 1296 60
NM_007041.3 NP_008972.2 7 - 8 1061 60
XM_005269458.2 XP_005269515.1 8 - 9 1202 60
XM_005269459.3 XP_005269516.1 8 - 9 1425 60
XM_017015600.1 XP_016871089.1 7 - 8 924 60
XM_017015602.1 XP_016871091.1 6 - 7 841 60
GenBank mRNA AF079099.1 - 7 - 8 935 60
AK299101.1 - 7 - 8 947 60
AK303604.1 - 5 - 6 892 60
AK313057.1 - 7 - 8 1021 60
BC022026.1 - 7 - 8 1034 4825006 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012056.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001713.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATE1

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

protein arginylation
arginyltransferase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 ENST00000224652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 ENST00000369040.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 ENST00000369043.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 ENST00000470613.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 ENST00000535655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 ENST00000540606.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 ENST00000543447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 NM_001001976.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 NM_007041.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022660.hg.1 TC10001713.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000224652.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000369040.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000369043.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000423243.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000481784.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000540606.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000543447.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001001976.2 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001288734.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001288735.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001288736.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_007041.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NR_110006.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000224652.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000369040.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000369043.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000423243.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000481784.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000540606.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 ENST00000543447.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001001976.2 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001288734.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001288735.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_001288736.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NM_007041.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 NR_110006.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182479.hg.1 TC1000012056.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0