This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01023852_m1
See other NUDT6 GE Assays ›
Gene Symbol:
NUDT6
Gene Name:
nudix hydrolase 6
Gene Aliases:
ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1
Chromosome Location:
Chr.4: 122892644 - 122923004 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01023852_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007083.4 NP_009014.2 1 - 2 273 59
GenBank mRNA AF019633.2 - 1 - 2 273 59
AK291871.1 - 1 - 2 272 59
BC009842.2 - 1 - 2 253 3942416 59
BG386740.1 - 1 - 2 251 59
CB128685.1 - 1 - 2 274 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400012961.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001523.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NUDT6

Panther Classification:

Molecular Function -

nucleotide phosphatase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
growth factor activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04021630.hg.1 ENST00000304430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021630.hg.1 ENST00000339154.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021630.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04021630.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04021630.hg.1 TC04001523.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04021634.hg.1 ENST00000304430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021634.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04021634.hg.1 TC04001523.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04021635.hg.1 ENST00000304430.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021635.hg.1 NM_007083.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04021635.hg.1 TC04001523.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000502270.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay TC0400012961.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000510735.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay PSR0400190948.hg.1 TC0400012961.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000339154.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 NM_198041.2 Refseq
Affymetrix Human Clariom D Assay PSR0400190944.hg.1 TC0400012961.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000304430.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000503370.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000512116.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 ENST00000513517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 NM_007083.4 Refseq
Affymetrix Human Clariom D Assay PSR0400190949.hg.1 TC0400012961.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0