This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01017345_m1
See other SLC25A18 GE Assays ›
Gene Symbol:
SLC25A18
Gene Name:
solute carrier family 25 member 18
Gene Aliases:
GC2
Chromosome Location:
Chr.22: 17562470 - 17590890 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01017345_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC25A18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001303484.1 NP_001290413.1 4 - 5 675 61
NM_031481.2 NP_113669.1 5 - 6 738 61
XM_011546149.2 XP_011544451.1 5 - 6 1667 61
XM_011546150.2 XP_011544452.1 5 - 6 1537 61
XM_011546151.2 XP_011544453.1 5 - 6 926 61
XM_011546152.1 XP_011544454.1 5 - 6 553 61
XM_011546153.2 XP_011544455.1 5 - 6 1548 61
XM_011546154.2 XP_011544456.1 5 - 6 795 61
XM_017028968.1 XP_016884457.1 5 - 6 421 61
XM_017028969.1 XP_016884458.1 5 - 6 2567 61
XM_017028970.1 XP_016884459.1 6 - 7 2058 61
XM_017028971.1 XP_016884460.1 6 - 7 2047 61
XM_017028972.1 XP_016884461.1 6 - 7 1970 61
XM_017028973.1 XP_016884462.1 5 - 6 1995 61
XM_017028974.1 XP_016884463.1 6 - 7 1926 61
XM_017028975.1 XP_016884464.1 6 - 7 1838 61
GenBank mRNA AJ428203.1 - 5 - 6 694 61
AK309110.1 - 4 - 5 522 61
AY008285.1 - 5 - 6 694 61
BC031644.1 - 4 - 5 630 5179038 61
CR456578.1 - 3 - 4 324 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200006520.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000024.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC25A18

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

translation
transmembrane transport
structural constituent of ribosome
protein binding
symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22000159.hg.1 ENST00000327451.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22000159.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22000159.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22000159.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22000159.hg.1 TC22000024.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000467228.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000469889.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000496051.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay TC2200006520.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 ENST00000327451.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 ENST00000399813.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 ENST00000497401.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 NM_001303484.1 Refseq
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 NM_031481.2 Refseq
Affymetrix Human Clariom D Assay PSR2200144563.hg.1 TC2200006520.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0