Assay Details
Gene Symbol: PRKAR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321982.1 | NP_001308911.1 | 7 - 8 | 1057 | 100 | |
NM_001321983.1 | NP_001308912.1 | 7 - 8 | 1057 | 100 | ||
NM_001321989.1 | NP_001308918.1 | 6 - 7 | 973 | 100 | ||
NM_004157.3 | NP_004148.1 | 7 - 8 | 1057 | 100 | ||
XM_005265315.3 | XP_005265372.1 | 7 - 8 | 1042 | 100 | ||
XM_011533942.2 | XP_011532244.1 | 7 - 8 | 1042 | 100 | ||
XM_011533943.1 | XP_011532245.1 | 7 - 8 | 1042 | 100 | ||
XM_017006815.1 | XP_016862304.1 | 7 - 8 | 1042 | 100 | ||
XM_017006816.1 | XP_016862305.1 | 7 - 8 | 1042 | 100 | ||
GenBank mRNA | AK293042.1 | - | 7 - 8 | 1011 | 100 | |
BC002763.2 | - | 7 - 8 | 1035 | 3629579 | 100 | |
BT007225.1 | - | 7 - 8 | 803 | 100 | ||
KU178276.1 | - | 7 - 8 | 803 | 100 | ||
KU178277.1 | - | 7 - 8 | 803 | 100 | ||
X14968.1 | - | 7 - 8 | 992 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001389.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
blood coagulation
activation of protein kinase A activity
intracellular signal transduction
cellular response to glucagon stimulus
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
renal water homeostasis
blood coagulation
activation of protein kinase A activity
intracellular signal transduction
cellular response to glucagon stimulus
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03024490.hg.1 | ENST00000265563.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03024490.hg.1 | ENST00000296446.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03024490.hg.1 | ENST00000454963.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03024490.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03024490.hg.1 | TC03001389.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181303.hg.1 | TC0300011036.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
