This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01011279_m1
See other IWS1 GE Assays ›
Gene Symbol:
IWS1
Gene Name:
IWS1, SUPT6H interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.2: 127480807 - 127526886 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01011279_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IWS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017969.2 NP_060439.2 5 - 6 1771 93
XM_005263707.3 XP_005263764.1 5 - 6 1607 93
XM_006712626.3 XP_006712689.1 5 - 6 1685 93
XM_011511450.1 XP_011509752.1 5 - 6 1685 93
XM_017004467.1 XP_016859956.1 5 - 6 1772 93
XM_017004468.1 XP_016859957.1 5 - 6 1541 93
GenBank mRNA AK000868.1 - 5 - 6 1098 93
AK027561.1 - 5 - 6 1728 93
AK056881.1 - 5 - 6 772 93
AK294669.1 - 6 - 7 1479 93
AK296848.1 - 6 - 7 1579 93
AL834178.1 - 5 - 6 1719 93
BC017012.2 - 2 - 3 248 93
BC110536.1 - 5 - 6 1601 40035173 93
BC110537.1 - 5 - 6 1601 40035174 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200016727.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002281.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IWS1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
regulation of mRNA export from nucleus
regulation of mRNA processing
mRNA transport
regulation of histone H4 acetylation
regulation of histone H3-K36 trimethylation
DNA binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02036674.hg.1 ENST00000295321.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036674.hg.1 ENST00000433551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036674.hg.1 NM_017969.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036674.hg.1 TC02002281.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200227687.hg.1 ENST00000295321.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200227687.hg.1 NM_017969.2 Refseq
Affymetrix Human Clariom D Assay PSR0200227687.hg.1 ENST00000295321.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200227687.hg.1 NM_017969.2 Refseq
Affymetrix Human Clariom D Assay PSR0200227687.hg.1 TC0200016727.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0