This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01010988_g1
See other DUOX2 GE Assays ›
Gene Symbol:
DUOX2
Gene Name:
dual oxidase 2
Gene Aliases:
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
Chromosome Location:
Chr.15: 45092653 - 45114161 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01010988_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUOX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014080.4 NP_054799.4 22 - 23 3120 124
XM_005254421.2 XP_005254478.1 22 - 23 3120 124
GenBank mRNA AF181972.1 - 14 - 15 1903 124
AF230496.1 - 22 - 23 3102 124
AF267981.1 - 22 - 23 3120 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009281.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001305.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DUOX2

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

thyroid hormone generation
response to oxidative stress
fertilization
response to virus
cytokine-mediated signaling pathway
bone mineralization
thyroid gland development
multicellular organism growth
cuticle development
hormone biosynthetic process
hydrogen peroxide catabolic process
inner ear development
adenohypophysis morphogenesis
hydrogen peroxide biosynthetic process
response to cAMP
oxidation-reduction process
cellular oxidant detoxification
peroxidase activity
calcium ion binding
NAD(P)H oxidase activity
heme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15012476.hg.1 ENST00000389039.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15012476.hg.1 NM_014080.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15012476.hg.1 TC15001305.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 ENST00000389039.10 Ensembl
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 ENST00000558383.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 ENST00000603300.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 NM_014080.4 Refseq
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 ENST00000389039.10 Ensembl
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 ENST00000558383.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 ENST00000603300.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 NM_014080.4 Refseq
Affymetrix Human Clariom D Assay PSR1500167980.hg.1 TC1500009281.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0