This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01010663_g1
See other AARS GE Assays ›
Gene Symbol:
AARS
Gene Name:
alanyl-tRNA synthetase
Gene Aliases:
CMT2N, EIEE29
Chromosome Location:
Chr.16: 70252295 - 70289552 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01010663_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AARS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001605.2 NP_001596.2 3 - 4 442 65
GenBank mRNA AK222824.1 - 3 - 4 439 65
AK225116.1 - 3 - 4 439 65
AK299098.1 - 3 - 4 418 65
AK307782.1 - 3 - 4 439 65
BC011451.1 - 3 - 4 374 3450606 65
D32050.1 - 3 - 4 442 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600010752.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16001234.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AARS

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

hair follicle development
tRNA modification
tRNA aminoacylation for protein translation
alanyl-tRNA aminoacylation
regulation of translational fidelity
protein folding
tRNA processing
cerebellar Purkinje cell layer development
endoplasmic reticulum unfolded protein response
response to amino acid
negative regulation of neuron apoptotic process
neuromuscular process controlling balance
tRNA binding
aminoacyl-tRNA editing activity
alanine-tRNA ligase activity
ATP binding
zinc ion binding
amino acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16017047.hg.1 ENST00000261772.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16017047.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16017047.hg.1 TC16001234.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000567490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000567490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000567490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay PSR1600183125.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay PSR1600183125.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay PSR1600183125.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay PSR1600183125.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay PSR1600183125.hg.1 TC1600010752.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000261772.12 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000564359.5 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000565361.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000566969.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000567490.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000567490.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000567490.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569790.2 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 ENST00000569825.1 Ensembl
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq
Affymetrix Human Clariom D Assay TC1600010752.hg.1 NM_001605.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0