This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01009406_s1
See other LINC01023 GE Assays ›
Gene Symbol:
LINC01023
Gene Name:
long intergenic non-protein coding RNA 1023
Gene Aliases:
HP07349
Chromosome Location:
Chr.5: 108727825 - 108728261 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01009406_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01023
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK126449.1 - 2 1378 140
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX117545.1 - 1 353 Chr. 5 - 108726365 - 108727124 [-] on Build GRCh38 140

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream F-box and leucine rich repeat protein 17 FBXL17 FBXO13,
Fbl17,
Fbx13
-343.6kb Sense Chr. 5 - 107859033 - 108382098 on NCBI Build GRCh38
Downstream FER tyrosine kinase FER PPP1R74,
TYK3,
p94-Fer
18.1kb Anti-sense Chr. 5 - 108746085 - 109196841 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 230243 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0