This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01006988_g1
See other FKBP10 GE Assays ›
Gene Symbol:
FKBP10
Gene Name:
FK506 binding protein 10
Gene Aliases:
BRKS1, FKBP65, OI11, OI6, PPIASE, hFKBP65
Chromosome Location:
Chr.17: 41812262 - 41823217 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01006988_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FKBP10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK025874.1 - 2 - 3 255 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000321562.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000429461.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000455106.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000464180.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000487489.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000489591.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000490938.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585664.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 ENST00000585922.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq
Affymetrix Human Clariom D Assay TC1700007858.hg.1 NM_021939.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0