This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00996837_m1
See other SLC18A2 GE Assays ›
Gene Symbol:
SLC18A2
Gene Name:
solute carrier family 18 member A2
Gene Aliases:
SVAT, SVMT, VAT2, VMAT2
Chromosome Location:
Chr.10: 117241073 - 117279430 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00996837_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC18A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003054.4 NP_003045.2 2 - 3 285 99
GenBank mRNA AK314997.1 - 2 - 3 238 99
BC030593.1 - 2 - 3 270 99
BC108927.1 - 1 - 2 150 40025174 99
BC108928.2 - 1 - 2 150 40025175 99
DB458870.1 - 2 - 3 285 99
L09118.1 - 2 - 3 227 99
L14269.1 - 2 - 3 155 99
L23205.1 - 2 - 3 235 99
X71354.1 - 2 - 3 185 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009017.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000850.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC18A2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

response to amphetamine
serotonin transport
chemical synaptic transmission
neurotransmitter secretion
aging
locomotory behavior
response to herbicide
post-embryonic development
response to zinc ion
aminergic neurotransmitter loading into synaptic vesicle
monoamine transport
dopamine transport
insulin secretion
endocytic recycling
cellular response to drug
sequestering of neurotransmitter
response to cocaine
glucose homeostasis
response to starvation
response to corticosterone
negative regulation of neurotransmitter transport
transmembrane transport
cellular response to ammonium ion
neurotransmitter loading into synaptic vesicle
negative regulation of reactive oxygen species biosynthetic process
amine transmembrane transporter activity
drug binding
monoamine transmembrane transporter activity
serotonin transmembrane transporter activity
enzyme binding
heat shock protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010315.hg.1 ENST00000298472.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010315.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010315.hg.1 NM_003054.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010315.hg.1 TC10000850.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000161784.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000161784.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000161784.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay PSR1000161784.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000161784.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000161784.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay PSR1000161784.hg.1 TC1000009017.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000298472.9 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 ENST00000497497.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq
Affymetrix Human Clariom D Assay TC1000009017.hg.1 NM_003054.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0