This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00995004_m1
See other METAP1D GE Assays ›
Gene Symbol:
METAP1D
Gene Name:
methionyl aminopeptidase type 1D (mitochondrial)
Gene Aliases:
MAP 1D, MAP1D, MetAP 1D, Metap1l
Chromosome Location:
Chr.2: 171999897 - 172082430 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00995004_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METAP1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322279.1 NP_001309208.1 3 - 4 470 85
NM_199227.2 NP_954697.1 3 - 4 420 85
XM_017003750.1 XP_016859239.1 3 - 4 419 85
GenBank mRNA AY374142.1 - 3 - 4 420 85
BC113644.1 - 3 - 4 365 8322696 85
DQ005576.1 - 3 - 4 734 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009966.hg
This assay targets the same location as HTA transcript cluster(s) TC02001025.hg

Additional Information:

This assay is in the 5'UTR region of the NM_001322279.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METAP1D

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
peptidyl-methionine modification
N-terminal protein amino acid modification
protein initiator methionine removal
aminopeptidase activity
metalloexopeptidase activity
metal ion binding
metalloaminopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02015238.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02015238.hg.1 NM_199227.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02015238.hg.1 TC02001025.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200169912.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200169912.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200169912.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay PSR0200169912.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200169912.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Clariom D Assay PSR0200169912.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay PSR0200169912.hg.1 TC0200009966.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000315796.4 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000392582.2 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000468537.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000488581.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000491440.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493035.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 ENST00000493742.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq
Affymetrix Human Clariom D Assay TC0200009966.hg.1 NM_199227.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0