This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00982629_m1
See other POLH GE Assays ›
Gene Symbol:
POLH
Gene Name:
DNA polymerase eta
Gene Aliases:
RAD30, RAD30A, XP-V, XPV
Chromosome Location:
Chr.6: 43576141 - 43620523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00982629_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291970.1 NP_001278899.1 4 - 5 799 64
NM_006502.2 NP_006493.1 4 - 5 799 64
XM_005249186.3 XP_005249243.1 3 - 4 652 64
GenBank mRNA AB024313.1 - 4 - 5 732 64
AF158185.1 - 4 - 5 660 64
AJ420484.1 - 1 - 2 116 64
AK023893.1 - 4 - 5 790 64
AK025631.1 - 127 64
AK122807.1 - 1 - 2 707 64
AK294430.1 - 4 - 5 780 64
AK316151.1 - 3 - 4 614 64
BC015742.1 - 4 - 5 643 4872554 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008099.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000605.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLH

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA synthesis involved in DNA repair
DNA replication
DNA repair
regulation of DNA repair
pyrimidine dimer repair
response to UV-C
translesion synthesis
error-free translesion synthesis
cellular response to UV-C
damaged DNA binding
DNA-directed DNA polymerase activity
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06007238.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007238.hg.1 ENST00000372236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007238.hg.1 ENST00000535400.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007238.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007238.hg.1 TC06000605.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000443535.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000443535.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 ENST00000443535.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007237.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007237.hg.1 ENST00000372236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007237.hg.1 ENST00000535400.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007237.hg.1 NM_006502.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007237.hg.1 TC06000605.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000443535.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000443535.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 ENST00000443535.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay TC0600008099.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay PSR0600156592.hg.1 TC0600008099.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 ENST00000372226.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 ENST00000372236.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 NM_001291969.1 Refseq
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 NM_001291970.1 Refseq
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 NM_006502.2 Refseq
Affymetrix Human Clariom D Assay PSR0600156593.hg.1 TC0600008099.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0