This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00980085_g1
See other ERN1 GE Assays ›
Gene Symbol:
ERN1
Gene Name:
endoplasmic reticulum to nucleus signaling 1
Gene Aliases:
IRE1, IRE1P, IRE1a, hIRE1p
Chromosome Location:
Chr.17: 64039142 - 64130819 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00980085_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001433.3 NP_001424.3 14 - 15 1874 54
XM_017024347.1 XP_016879836.1 14 - 15 3228 54
XM_017024348.1 XP_016879837.1 13 - 14 3126 54
GenBank mRNA AB209869.1 - 14 - 15 1857 54
AF059198.1 - 14 - 15 1857 54
AK292403.1 - 14 - 15 1900 54
BC130405.1 - 14 - 15 1807 40146436 54
BC130407.1 - 14 - 15 1807 40146438 54

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700011436.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001796.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ERN1

Panther Classification:

Molecular Function -

endoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

endothelial cell proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA cleavage
mRNA catabolic process
protein phosphorylation
activation of signaling protein activity involved in unfolded protein response
cell cycle arrest
activation of JUN kinase activity
regulation of macroautophagy
positive regulation of RNA splicing
response to endoplasmic reticulum stress
cellular response to vascular endothelial growth factor stimulus
peptidyl-serine autophosphorylation
IRE1-mediated unfolded protein response
protein autophosphorylation
mRNA splicing, via endonucleolytic cleavage and ligation
mRNA endonucleolytic cleavage involved in unfolded protein response
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to glucose stimulus
RNA phosphodiester bond hydrolysis, endonucleolytic
positive regulation of endoplasmic reticulum unfolded protein response
insulin metabolic process
peptidyl-serine trans-autophosphorylation
magnesium ion binding
endoribonuclease activity
protein serine/threonine kinase activity
protein binding
ATP binding
enzyme binding
Hsp70 protein binding
identical protein binding
protein homodimerization activity
ADP binding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000433197.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000577567.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000579249.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583028.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583077.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583077.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583077.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000583896.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000584041.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 ENST00000606895.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011436.hg.1 NM_001433.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0