This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979877_m1
See other CLASP2 GE Assays ›
Gene Symbol:
CLASP2
Gene Name:
cytoplasmic linker associated protein 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 33496245 - 33718356 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00979877_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLASP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001207044.1 NP_001193973.1 12 - 13 1674 106
NM_015097.2 NP_055912.2 18 - 19 2135 106
XM_006713040.1 XP_006713103.1 18 - 19 2278 106
XM_006713041.1 XP_006713104.1 18 - 19 2275 106
XM_006713042.1 XP_006713105.1 18 - 19 2278 106
XM_006713048.1 XP_006713111.1 18 - 19 2278 106
XM_006713049.1 XP_006713112.1 18 - 19 2275 106
XM_006713050.1 XP_006713113.1 18 - 19 2278 106
XM_006713052.1 XP_006713115.1 18 - 19 2275 106
XM_006713053.1 XP_006713116.1 18 - 19 2278 106
XM_017005947.1 XP_016861436.1 18 - 19 2275 106
XM_017005948.1 XP_016861437.1 18 - 19 2275 106
XM_017005950.1 XP_016861439.1 18 - 19 2278 106
XM_017005951.1 XP_016861440.1 18 - 19 2275 106
XM_017005952.1 XP_016861441.1 18 - 19 2278 106
XM_017005953.1 XP_016861442.1 18 - 19 2275 106
XM_017005954.1 XP_016861443.1 18 - 19 2275 106
XM_017005955.1 XP_016861444.1 18 - 19 2278 106
XM_017005956.1 XP_016861445.1 18 - 19 2278 106
XM_017005957.1 XP_016861446.1 18 - 19 2278 106
XM_017005958.1 XP_016861447.1 18 - 19 2278 106
XM_017005959.1 XP_016861448.1 18 - 19 2275 106
XM_017005962.1 XP_016861451.1 18 - 19 2275 106
XM_017005963.1 XP_016861452.1 18 - 19 2278 106
XM_017005964.1 XP_016861453.1 18 - 19 2275 106
XM_017005968.1 XP_016861457.1 12 - 13 1418 106
XM_017005969.1 XP_016861458.1 12 - 13 1428 106
XM_017005970.1 XP_016861459.1 12 - 13 1677 106
XM_017005971.1 XP_016861460.1 12 - 13 1428 106
XM_017005972.1 XP_016861461.1 12 - 13 2005 106
XM_017005973.1 XP_016861462.1 12 - 13 1674 106
XM_017005974.1 XP_016861463.1 12 - 13 1674 106
XM_017005975.1 XP_016861464.1 12 - 13 1418 106
XM_017005977.1 XP_016861466.1 12 - 13 1677 106
XM_017005978.1 XP_016861467.1 12 - 13 1418 106
XM_017005979.1 XP_016861468.1 12 - 13 1428 106
XM_017005980.1 XP_016861469.1 12 - 13 2005 106
XM_017005981.1 XP_016861470.1 12 - 13 1418 106
XM_017005982.1 XP_016861471.1 12 - 13 1428 106
XM_017005983.1 XP_016861472.1 12 - 13 1677 106
XM_017005984.1 XP_016861473.1 12 - 13 1674 106
XM_017005985.1 XP_016861474.1 12 - 13 2005 106
GenBank mRNA AB014527.2 - 12 - 13 1313 106
AK091451.1 - 18 - 19 2275 106
AK297322.1 - 11 - 12 1337 106
BC140778.1 - 18 - 19 1971 9021646 106

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLASP2

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
microtubule organizing center organization
regulation of microtubule polymerization or depolymerization
regulation of microtubule-based process
microtubule anchoring
positive regulation of exocytosis
cell division
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
establishment of protein localization to plasma membrane
positive regulation of extracellular matrix disassembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
microtubule plus-end binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0