This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979694_g1
See other SMG1 GE Assays ›
Gene Symbol:
SMG1
Gene Name:
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Gene Aliases:
61E3.4, ATX, LIP
Chromosome Location:
Chr.16: 18804853 - 18926417 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00979694_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015092.4 NP_055907.3 44 - 45 7552 70
XM_005255182.3 XP_005255239.1 43 - 44 7649 70
XM_005255183.3 XP_005255240.1 44 - 45 7707 70
XM_005255184.3 XP_005255241.1 42 - 43 7493 70
XM_011545769.2 XP_011544071.1 43 - 44 7649 70
XM_011545770.1 XP_011544072.1 31 - 32 5446 70
XM_017023066.1 XP_016878555.1 43 - 44 7459 70
XM_017023067.1 XP_016878556.1 31 - 32 5509 70
GenBank mRNA AB007881.2 - 12 - 13 2170 70
AB061371.1 - 45 - 46 7504 70
AF186377.1 - 19 - 20 3533 70
AF395444.1 - 42 - 43 6859 70
AY014957.1 - 32 - 33 5431 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009620.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000916.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMG1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
telomere maintenance
DNA repair
mRNA export from nucleus
response to stress
peptidyl-serine phosphorylation
protein autophosphorylation
phosphatidylinositol phosphorylation
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
telomeric DNA binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16013142.hg.1 ENST00000389467.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16013142.hg.1 ENST00000446231.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16013142.hg.1 NM_015092.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16013142.hg.1 TC16000916.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600173597.hg.1 ENST00000446231.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600173597.hg.1 ENST00000565324.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600173597.hg.1 NM_015092.4 Refseq
Affymetrix Human Clariom D Assay PSR1600173597.hg.1 ENST00000446231.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600173597.hg.1 ENST00000565324.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600173597.hg.1 NM_015092.4 Refseq
Affymetrix Human Clariom D Assay PSR1600173597.hg.1 TC1600009620.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0