This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979578_m1
See other ABCA4 GE Assays ›
Gene Symbol:
ABCA4
Gene Name:
ATP binding cassette subfamily A member 4
Gene Aliases:
ABC10, ABCR, ARMD2, CORD3, FFM, RMP, RP19, STGD, STGD1
Chromosome Location:
Chr.1: 93992837 - 94121149 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00979578_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000350.2 NP_000341.2 26 - 27 3973 93
GenBank mRNA AB210040.1 - 26 - 27 3959 93
AF000148.1 - 26 - 27 3956 93
AF001945.1 - 26 - 27 3950 93
CR627391.1 - 4 - 5 385 93
U88667.1 - 26 - 27 3950 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014977.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002883.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABCA4

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
phospholipid transfer to membrane
transport
lipid transport
visual perception
phototransduction, visible light
phospholipid translocation
photoreceptor cell maintenance
transmembrane transport
phospholipid-translocating ATPase activity
transporter activity
eye pigment precursor transporter activity
ATP binding
ATPase activity, coupled to transmembrane movement of substances

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01045002.hg.1 ENST00000370225.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01045002.hg.1 ENST00000546054.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01045002.hg.1 NM_000350.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01045002.hg.1 TC01002883.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100211584.hg.1 ENST00000370225.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100211584.hg.1 ENST00000536513.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211584.hg.1 NM_000350.2 Refseq
Affymetrix Human Clariom D Assay PSR0100211584.hg.1 ENST00000370225.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100211584.hg.1 ENST00000536513.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211584.hg.1 NM_000350.2 Refseq
Affymetrix Human Clariom D Assay PSR0100211584.hg.1 TC0100014977.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0