Assay Details
Gene Symbol: MED12L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053002.5 | NP_443728.3 | 34 - 35 | 5110 | 101 | |
XM_006713487.3 | XP_006713550.1 | 36 - 37 | 5487 | 101 | ||
XM_011512390.2 | XP_011510692.1 | 35 - 36 | 5382 | 101 | ||
XM_011512394.2 | XP_011510696.1 | 36 - 37 | 5487 | 101 | ||
XM_017005676.1 | XP_016861165.1 | 36 - 37 | 5487 | 101 | ||
XM_017005677.1 | XP_016861166.1 | 36 - 37 | 5484 | 101 | ||
XM_017005678.1 | XP_016861167.1 | 36 - 37 | 5487 | 101 | ||
XM_017005679.1 | XP_016861168.1 | 35 - 36 | 5217 | 101 | ||
XM_017005680.1 | XP_016861169.1 | 34 - 35 | 5205 | 101 | ||
XM_017005681.1 | XP_016861170.1 | 21 - 22 | 2873 | 101 | ||
GenBank mRNA | AB046855.1 | - | 21 - 22 | 2853 | 101 | |
AF399708.1 | - | 34 - 35 | 5019 | 101 | ||
AK022714.1 | - | 18 - 19 | 2559 | 101 | ||
AK128165.1 | - | 18 - 19 | 2699 | 101 | ||
BC144559.1 | - | 32 - 33 | 4594 | 9053087 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000363124.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
