This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00971313_m1
See other CSAD GE Assays ›
Gene Symbol:
CSAD
Gene Name:
cysteine sulfinic acid decarboxylase
Gene Aliases:
CSD, PCAP
Chromosome Location:
Chr.12: 53157663 - 53180909 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00971313_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSAD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244705.1 NP_001231634.1 8 - 9 836 58
NM_015989.4 NP_057073.4 8 - 9 933 58
XM_011538446.2 XP_011536748.1 7 - 8 1687 58
XM_017019417.1 XP_016874906.1 4 - 5 413 58
GenBank mRNA AF116547.1 - 7 - 8 644 58
AF116548.1 - 8 - 9 883 58
AK315265.1 - 8 - 9 860 58
BC052249.1 - 8 - 9 836 58
BC098278.1 - 7 - 8 709 40006057 58
BC098342.1 - 7 - 8 709 40006051 58
BC099717.1 - 7 - 8 709 40006058 58
BC105918.1 - 7 - 8 709 40006053 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200010793.hg
This assay targets the same location as HTA transcript cluster(s) TC12001543.hg

Additional Information:

This assay is in the 5'UTR region of the XM_017019417.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CSAD

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

carboxylic acid metabolic process
taurine biosynthetic process
sulfinoalanine decarboxylase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0