This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00970034_g1
See other ASNA1 GE Assays ›
Gene Symbol:
ASNA1
Gene Name:
arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)
Gene Aliases:
ARSA-I, ARSA1, ASNA-I, GET3, TRC40
Chromosome Location:
Chr.19: 12737492 - 12748323 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00970034_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASNA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF047469.1 - 3 - 4 461 68
AK223363.1 - 3 - 4 468 68
AK291855.1 - 3 - 4 462 68
AK293588.1 - 402 68
BC002651.2 - 3 - 4 454 3609717 68
BM718941.1 - 3 - 4 509 68
U60276.1 - 3 - 4 405 68

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transporter

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150936.hg.1 TC1900007089.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000590633.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150938.hg.1 TC1900007089.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000357332.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000586561.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000591090.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 ENST00000611688.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 NM_004317.3 Refseq
Affymetrix Human Clariom D Assay PSR1900150939.hg.1 TC1900007089.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0