This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00961614_s1
See other TDRD15 GE Assays ›
Gene Symbol:
TDRD15
Gene Name:
tudor domain containing 15
Gene Aliases:
-
Chromosome Location:
Chr.2: 21123932 - 21147325 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00961614_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TDRD15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306137.1 NP_001293066.1 4 6351 110
XM_011533212.1 XP_011531514.1 4 6446 110
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF113010.1 - 1 Chr. 2 - 21142922 - 21144312 [-] on Build GRCh38 110

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream apolipoprotein B APOB FLDB,
LDLCQ4,
apoB-100,
apoB-48
-93.4kb Anti-sense Chr. 2 - 21001429 - 21044073 on NCBI Build GRCh38
Downstream uncharacterized LOC645949 LOC645949 543.1kb Anti-sense Chr. 2 - 21687434 - 21710652 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0