Assay Details
Gene Symbol: CD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014207.3 | NP_055022.2 | 1 - 2 | 160 | 80 | |
GenBank mRNA | AK292698.1 | - | 1 - 2 | 129 | 80 | |
BC027901.1 | - | 1 - 2 | 123 | 5228913 | 80 | |
DB143341.1 | - | 1 - 2 | 160 | 80 | ||
X04391.1 | - | 1 - 2 | 129 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000526.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11006282.hg.1 | ENST00000347785.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11006282.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11006282.hg.1 | TC11000526.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11006283.hg.1 | ENST00000347785.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11006283.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11006283.hg.1 | TC11000526.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11006284.hg.1 | ENST00000347785.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11006284.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11006284.hg.1 | TC11000526.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007790.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155166.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155166.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155166.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155166.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155166.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155166.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155166.hg.1 | TC1100007790.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100155167.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155167.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155167.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155167.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155167.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155167.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155167.hg.1 | TC1100007790.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100155164.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155164.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155164.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155164.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155164.hg.1 | TC1100007790.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100155169.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155169.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155169.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155169.hg.1 | ENST00000347785.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155169.hg.1 | ENST00000544014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100155169.hg.1 | NM_014207.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100155169.hg.1 | TC1100007790.hg.1 | Clariom D TC |
