This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00960479_g1
See other TM9SF3 GE Assays ›
Gene Symbol:
TM9SF3
Gene Name:
transmembrane 9 superfamily member 3
Gene Aliases:
EP70-P-iso, SMBP
Chromosome Location:
Chr.10: 96518110 - 96587052 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00960479_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TM9SF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020123.3 NP_064508.3 15 2355 251
XM_011539976.2 XP_011538278.1 15 2435 251
XM_011539977.1 XP_011538279.1 15 2484 251
GenBank mRNA AF160213.1 - 15 2148 251
AF269150.1 - 15 2355 251
AK000756.1 - 13 1758 251
AK027438.1 - 10 1405 251
AK027788.1 - 14 2011 251
AK074824.1 - 12 1646 250
AK075093.1 - 10 1393 251
AK075451.1 - 4 758 251
AY358819.1 - 14 2024 251
BC020959.1 - 14 1935 251
BC136788.1 - 15 2355 9020778 251
BC136789.1 - 15 2355 9020779 251

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011522.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001558.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TM9SF3

Panther Classification:

Molecular Function -

transporter

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10019760.hg.1 ENST00000371142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10019760.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10019760.hg.1 TC10001558.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10019761.hg.1 ENST00000371142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10019761.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10019761.hg.1 TC10001558.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000475401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000475401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000475401.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay PSR1000178319.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000178319.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay PSR1000178319.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000178319.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay PSR1000178319.hg.1 TC1000011522.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000371142.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000443638.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000464654.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000475401.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000475401.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000475401.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000485093.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 ENST00000490192.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq
Affymetrix Human Clariom D Assay TC1000011522.hg.1 NM_020123.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0