This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00955720_m1
See other VSIG10 GE Assays ›
Gene Symbol:
VSIG10
Gene Name:
V-set and immunoglobulin domain containing 10
Gene Aliases:
-
Chromosome Location:
Chr.12: 118063593 - 118104402 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00955720_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,681.00
Your Price
Online offer:
2681.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019086.5 NP_061959.2 7 - 8 1677 68
XM_005253908.4 XP_005253965.1 6 - 7 1030 68
XM_006719467.2 XP_006719530.1 6 - 7 1374 68
XM_006719468.2 XP_006719531.1 5 - 6 769 68
XM_017019504.1 XP_016874993.1 8 - 9 1691 68
XM_017019505.1 XP_016874994.1 7 - 8 1297 68
XM_017019506.1 XP_016874995.1 7 - 8 1811 68
GenBank mRNA AK000681.1 - 7 - 8 1707 68
BC028698.2 - 7 - 8 1621 4838234 68
BC034471.2 - 7 - 8 1616 4825667 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200012855.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12002017.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VSIG10

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12026500.hg.1 ENST00000359236.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026500.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12026500.hg.1 TC12002017.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000540117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000540117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000545722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000545722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay PSR1200204545.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay PSR1200204545.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay PSR1200204545.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay PSR1200204545.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay PSR1200204545.hg.1 TC1200012855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000359236.9 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000536905.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000538357.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000540117.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000540117.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000541855.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000542195.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000545259.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000545722.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000545722.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 ENST00000612851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Affymetrix Human Clariom D Assay TC1200012855.hg.1 NM_019086.5 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 103271 - -
ABI Human Genome Survey Microarray Version 2.0 195597 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0