This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00950504_g1
See other GIF GE Assays ›
Gene Symbol:
GIF
Gene Name:
gastric intrinsic factor
Gene Aliases:
IF, IFMH, INF, TCN3
Chromosome Location:
Chr.11: 59829268 - 59845529 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00950504_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GIF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005142.2 NP_005133.2 3 - 4 420 131
XM_011544939.2 XP_011543241.1 3 - 4 448 131
GenBank mRNA AK314275.1 - 3 - 4 439 131
BC037958.1 - 3 - 4 420 5759934 131
X76562.1 - 3 - 4 419 131

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100010991.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001836.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GIF

Gene Ontology Categories:

Function(s) Process(es)

cobalt ion transport
cobalamin metabolic process
cobalamin transport
cobalamin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000257248.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000525058.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000532070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 ENST00000533847.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010991.hg.1 NM_005142.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0