This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00947069_g1
See other MNT GE Assays ›
Gene Symbol:
MNT
Gene Name:
MAX network transcriptional repressor
Gene Aliases:
MAD6, MXD6, ROX, bHLHd3
Chromosome Location:
Chr.17: 2384060 - 2400964 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00947069_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MNT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020310.2 NP_064706.1 3 - 4 943 75
XM_011523868.2 XP_011522170.1 3 - 4 880 75
GenBank mRNA AK291596.1 - 3 - 4 1040 75
BC117563.1 - 3 - 4 693 3904263 75
X96401.1 - 3 - 4 903 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700009436.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000994.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MNT

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
multicellular organism development
cell aging
negative regulation of cell proliferation
regulation of cell cycle
negative regulation of apoptotic signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17013271.hg.1 ENST00000174618.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17013271.hg.1 ENST00000404961.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17013271.hg.1 NM_020310.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17013271.hg.1 TC17000994.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700172236.hg.1 ENST00000174618.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700172236.hg.1 NM_020310.2 Refseq
Affymetrix Human Clariom D Assay PSR1700172236.hg.1 ENST00000174618.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700172236.hg.1 NM_020310.2 Refseq
Affymetrix Human Clariom D Assay PSR1700172236.hg.1 TC1700009436.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0