This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00945166_g1
See other POLG2 GE Assays ›
Gene Symbol:
POLG2
Gene Name:
DNA polymerase gamma 2, accessory subunit
Gene Aliases:
HP55, MTPOLB, PEOA4, POLB, POLG-BETA, POLGB
Chromosome Location:
Chr.17: 64477785 - 64497066 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00945166_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLG2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007215.3 NP_009146.2 2 - 3 789 122
XM_006721651.2 XP_006721714.1 2 - 3 742 122
XM_017024072.1 XP_016879561.1 2 - 3 742 122
GenBank mRNA AF142992.1 - 2 - 3 772 122
AF177201.1 - 2 - 3 691 122
AF184344.1 - 2 - 3 772 122
AK307801.1 - 2 - 3 742 122
BC000913.2 - 2 - 3 537 122
BC009194.2 - 2 - 3 759 3949617 122
U94703.1 - 2 - 3 773 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700011448.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001799.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLG2

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
DNA replication
DNA-dependent DNA replication
DNA repair
glycyl-tRNA aminoacylation
mitochondrion organization
respiratory electron transport chain
mitochondrial DNA metabolic process
mitochondrion morphogenesis
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
glycine-tRNA ligase activity
protein binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000539111.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000577506.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000577506.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000577506.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000577506.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000577506.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000577506.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000577506.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578997.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578997.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578997.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578997.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000578997.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000580893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000581355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000582501.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 ENST00000585141.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011448.hg.1 NM_007215.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0