This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00940488_m1
See other SLC37A1 GE Assays ›
Gene Symbol:
SLC37A1
Gene Name:
solute carrier family 37 member 1
Gene Aliases:
G3PP
Chromosome Location:
Chr.21: 42499632 - 42581440 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00940488_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC37A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320537.1 NP_001307466.1 10 - 11 1889 100
NM_018964.3 NP_061837.3 11 - 12 1258 100
XM_011529614.2 XP_011527916.1 9 - 10 2030 100
XM_011529615.1 XP_011527917.1 9 - 10 2030 100
XM_017028376.1 XP_016883865.1 9 - 10 2030 100
XM_017028377.1 XP_016883866.1 11 - 12 1183 100
XM_017028378.1 XP_016883867.1 11 - 12 1642 100
XM_017028379.1 XP_016883868.1 12 - 13 1289 100
XM_017028380.1 XP_016883869.1 10 - 11 1089 100
XM_017028381.1 XP_016883870.1 11 - 12 1183 100
XM_017028382.1 XP_016883871.1 9 - 10 2371 100
XM_017028383.1 XP_016883872.1 11 - 12 1114 100
GenBank mRNA AF311320.1 - 10 - 11 1828 100
AJ269529.2 - 11 - 12 1258 100
BI912541.1 - 3 - 4 201 100
BM550728.1 - 3 - 4 290 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100008527.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000202.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC37A1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate transport
glucose-6-phosphate transport
phosphate ion transmembrane transport
glucose 6-phosphate:inorganic phosphate antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21002107.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002107.hg.1 ENST00000398341.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002107.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21002107.hg.1 TC21000202.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000484887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000484887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000484887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000487951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000487951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000487951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000398343.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000471277.5 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000484887.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000484887.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000484887.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000487951.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000487951.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000487951.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 ENST00000496416.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay TC2100008527.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay PSR2100158346.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay PSR2100158346.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100158346.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay PSR2100158346.hg.1 ENST00000352133.2 Ensembl
Affymetrix Human Clariom D Assay PSR2100158346.hg.1 ENST00000398341.7 Ensembl
Affymetrix Human Clariom D Assay PSR2100158346.hg.1 NM_018964.3 Refseq
Affymetrix Human Clariom D Assay PSR2100158346.hg.1 TC2100008527.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0