This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00934503_m1
See other WAC GE Assays ›
Gene Symbol:
WAC
Gene Name:
WW domain containing adaptor with coiled-coil
Gene Aliases:
BM-016, DESSH, PRO1741, Wwp4
Chromosome Location:
Chr.10: 28532426 - 28623112 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00934503_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WAC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_100264.2 NP_567822.1 2 - 3 461 137
XM_017016315.1 XP_016871804.1 2 - 3 281 137
XM_017016317.1 XP_016871806.1 2 - 3 281 137
XM_017016318.1 XP_016871807.1 2 - 3 281 137
GenBank mRNA AK055852.1 - 2 - 3 278 137
AK225846.1 - 2 - 3 274 137
BI463901.1 - 2 - 3 561 137

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK055852.1,NM_100264.2,XM_017016315.1,XM_017016317.1,XM_017016318.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene WAC

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
cellular response to DNA damage stimulus
histone monoubiquitination
positive regulation of macroautophagy
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
G1 DNA damage checkpoint
positive regulation of transcription, DNA-templated
histone H2B conserved C-terminal lysine ubiquitination
RNA polymerase II core binding
chromatin binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0