This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00932492_m1
See other GBA3 GE Assays ›
Gene Symbol:
GBA3
Gene Name:
glucosylceramidase beta 3 (gene/pseudogene)
Gene Aliases:
CBG, CBGL1, GLUC, KLRP
Chromosome Location:
Chr.4: 22692914 - 22819575 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00932492_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GBA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277225.1 NP_001264154.1 4 - 5 1204 102
NM_020973.4 NP_066024.1 4 - 5 1302 102
NR_102355.1 4 - 5 1302 102
NR_102357.1 4 - 5 1204 102
GenBank mRNA AB017913.1 - 4 - 5 1251 102
AF317840.1 - 4 - 5 1200 102
AF323990.1 - 4 - 5 1249 102
AK222963.1 - 4 - 5 1249 102
AK292746.1 - 4 - 5 1249 102
AK298377.1 - 4 - 5 1204 102
AK313189.1 - 4 - 5 1254 102
BC101829.1 - 4 - 5 1237 8069335 102
BC136264.1 - 4 - 5 1237 9020251 102
CB163648.1 - 2 - 3 276 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400007054.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000168.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GBA3

Panther Classification:

Molecular Function -

glycosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycosphingolipid metabolic process
glycoside catabolic process
glycosylceramide catabolic process
glucosylceramidase activity
beta-galactosidase activity
beta-glucosidase activity
glycosylceramidase activity
scopolin beta-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04002618.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002618.hg.1 ENST00000508166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002618.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002618.hg.1 ENST00000511446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002618.hg.1 TC04000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04002614.hg.1 ENST00000508166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002614.hg.1 ENST00000511446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002614.hg.1 TC04000168.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000504802.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000504802.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002616.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002616.hg.1 ENST00000508166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002616.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002616.hg.1 ENST00000511446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002616.hg.1 TC04000168.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148365.hg.1 TC0400007054.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000504802.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000504802.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay TC0400007054.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000503442.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000508166.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000508264.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000511446.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000610628.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 ENST00000613293.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 NR_102355.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 NR_102356.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 NR_102357.1 Refseq
Affymetrix Human Clariom D Assay PSR0400148367.hg.1 TC0400007054.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0