This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00909723_m1
See other SLC25A12 GE Assays ›
Gene Symbol:
SLC25A12
Gene Name:
solute carrier family 25 member 12
Gene Aliases:
AGC1, ARALAR
Chromosome Location:
Chr.2: 171783405 - 171894306 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00909723_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC25A12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003705.4 NP_003696.2 2 - 3 153 90
NR_047549.1 2 - 3 153 90
XM_011512070.2 XP_011510372.1 2 - 3 121 90
GenBank mRNA AJ496568.1 - 2 - 3 82 90
AK022828.1 - 2 - 3 91 90
AK091071.1 - 2 - 3 70 90
AK294638.1 - 2 - 3 91 90
BC016932.1 - 2 - 3 70 4420100 90
DN993435.1 - 2 - 3 155 90
Y14494.1 - 2 - 3 84 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK091071.1, XM_011512070.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC25A12

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
translation
aspartate transport
L-glutamate transport
malate-aspartate shuttle
response to calcium ion
L-glutamate transmembrane transport
L-aspartate transmembrane transport
structural constituent of ribosome
L-glutamate transmembrane transporter activity
calcium ion binding
acidic amino acid transmembrane transporter activity
L-aspartate transmembrane transporter activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 176097 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0